Learn More
ADRM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP1-30447
Product Code. 18727693
  • € 583.00 / 100µL
Description
ADRM1 Polyclonal specifically detects ADRM1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunoprecipitation
RUO
IgG
100 μL
Store at 4C. Do not freeze.