Learn More
ADRM1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-81769PCP
Product Code. 18696011
  • € 564.00 / 0.10mL
Description
ADRM1 Polyclonal antibody specifically detects ADRM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ADRM1
Polyclonal
PBS
0.1 mL
Mouse
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
PerCP
Peptide affinity purified