Learn More
Adropin Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25213-1-AP-150UL
 View more versions of this product
Product Code. 18059835
  • € 410.00 / 150µL
Description
The Adropin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Adropin. This antibody recognizes human antigen. The Adropin antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
Adropin
1000 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Immunofluorescence, Immunohistochemistry, ELISA
Adropin, C9orf165, energy homeostasis associated, ENHO, UNQ470
150 μL
375704
Store at -20°C. Stable for one year after shipment.