Learn More
Adropin Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25213-1-AP-20UL
This item is not returnable.
View return policy
Description
The Adropin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Adropin. This antibody recognizes human antigen. The Adropin antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| Adropin | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Immunofluorescence, Immunohistochemistry, ELISA | |
| Adropin, C9orf165, energy homeostasis associated, ENHO, UNQ470 | |
| 20 μL | |
| 375704 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |