missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADSSL1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13220709
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13220709 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13220709 Supplier Invitrogen™ Supplier No. PA525767

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine and porcine based on sequence homology.

ADSSL1 is a muscle isozyme of adenylosuccinate synthase (EC 6.3.4.4), which catalyzes the initial reaction in the conversion of inosine monophosphate (IMP) to adenosine monophosphate (AMP).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADSSL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADSSL1
Gene Accession No. P28650, Q8N142
Gene Alias adenylosuccinate synthase like 1; adenylosuccinate synthetase 1, muscle; adenylosuccinate synthetase isozyme 1; Adenylosuccinate synthetase like 1; adenylosuccinate synthetase, basic isozyme; Adenylosuccinate synthetase, muscle isozyme; Adenylosuccinate synthetase-like 1; Adss; adSS 1; Adss1; Adssl1; AI528595; AMPSase 1; IMP--aspartate ligase 1; M-type adenylosuccinate synthetase
Gene Symbols ADSSL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 96-123 amino acids from the N-terminal region of human ADSSL1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11565, 122622
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.