missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADTB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57886-25ul
This item is not returnable.
View return policy
Description
ADTB1 Polyclonal specifically detects ADTB1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ADTB1 | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Adapter-Related Protein Complex 1 Subunit Beta-1, Adaptor Protein Complex AP-1 Subunit Beta-1, Adaptor-Related Protein Complex 1, Beta 1 Subunit, AP-1 Complex Subunit Beta-1, AP105A, AP1B1 BAM22, beta1-adaptin, Beta-1-adaptin, Beta-Adaptin 1, beta-prime-adaptin, CLAPB2, Clathrin Assembly Protein Complex 1 Beta Large Chain, Golgi Adaptor HA1/AP1 Adaptin Beta Subunit, Plasma Membrane Adaptor HA2/AP2 Adaptor Beta Subunit | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
| AP1B1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GGLDSLMGDEPEGIGGTNFVAPPTAAVPANLGAPIGSGL | |
| 25 μL | |
| Membrane Vesicle Markers | |
| 162 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction