Learn More
Motic™ Phase Stop 0.3 Ph0 (4X)
Use phase contrast to improve visualization of transparent specimens. Motic™ Phase Stop 0.3 Ph0 (4X) is an annular light stop that fits in your condenser.
Brand: Motic™ 1101000202381
Additional Details : Weight : 0.01000kg
Description
- Boost contrast and detail discrimination without staining or fixing, thus permitting live cell study.
- Must be matched and aligned to the objective phase plate.
Specifications
Phase Stop Annulus | |
0.3 Ph0 (4X) | |
AE31E Microscope, 4X phase objective |