missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEBP2 Monoclonal Antibody (OTI10F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810263
This item is not returnable.
View return policy
Description
AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates ′Lys-9′ and ′Lys-27′ residues of histone H3.Specifications
| AEBP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6ZN18 | |
| AEBP2 | |
| Full length human recombit protein of human AEBP2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI10F3 | |
| Unconjugated | |
| AEBP2 | |
| AE binding protein 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 121536 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction