missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEBP2 Monoclonal Antibody (OTI8G9), TrueMAB™, OriGene
Description
AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates ′Lys-9′ and ′Lys-27′ residues of histone H3.
Specifications
Specifications
| Antigen | AEBP2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI8G9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | AEBP2 |
| Gene Accession No. | Q6ZN18 |
| Gene Alias | AE binding protein 2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?