missing translation for 'onlineSavingsMsg'
Learn More

AEBP2 Monoclonal Antibody (OTI8G9), TrueMAB™, OriGene

Product Code. p-200016384
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17593154 30 μL 30µL
17573154 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17593154 Supplier Origene Technologies Supplier No. TA810230S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AEBP2 Monoclonal antibody specifically detects AEBP2 in Human samples. It is validated for Western Blot

Specifications

Antigen AEBP2
Applications Western Blot
Classification Monoclonal
Clone OTI8G9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZFP37
Gene Accession No. Q9Y6Q3
Gene Alias adipocyte enhancer-binding protein 2; Adipocyte enhancer-binding protein 2 homolog; AE binding protein 2; AE(adipocyte enhancer)-binding protein 2; AE-binding protein 2; AE-binding protein 2 homolog; aebp2; AU023766; B230313N05Rik; LOW QUALITY PROTEIN: zinc finger protein AEBP2; si:dkey-158p11.2; wu:fk42f11; wu:fk52h12; zgc:63755; Zinc finger protein AEBP2
Host Species Mouse
Immunogen Full length human recombinant protein of human ZFP37 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7539
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.