missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AES Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5144208

Product Code. 17999652

  • € 378.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AES Polyclonal Antibody for IHC (P), ELISA

Adhesion to extracellular matrix regulates cell survival through both integrin engagement and appropriate cell spreading. Anoikis is the molecular mechanism of apop-tosis induced by integrin detachment. Amino-terminal enhancer of split (AES) is a member of the Groucho/ transducin-like enhancer of split (TLE) family of transcriptional regulators, a group of transcriptional co-repressors that play important roles in neurogenesis, segmentation, and sex determination. AES forms a complex with Bit1 (Bcl-2 inhibitor of transcription 1), a mitochondrial protein that is released into the cytoplasm upon onset of apoptosis. It has been suggested that this complex turns off a survival-promoting gene transcription program controlled by the TLE protein family. Interestingly, apoptosis of cells transfected with AES and Bit1 could be inhibited if the cells were allowed to attach to fibronectin through the alpha5beta1 integrin suggesting that the Bit1-AES pathway contributing to anoikis is regulated by integrins, and in particular, the alpha5beta1 integrin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AES
Polyclonal
Unconjugated
TLE5
Aes; AES-1; AES-2; AL024115; Amino enhancer of split; amino-terminal enhancer of split; Esp1; gp130-associated protein GAM; Grg; Grg5; Grg-5; Groucho-related protein 5; Protein ESP1; Protein GRG; related to Drosophila groucho; R-esp1; TLE family member 5; TLE family member 5, transcriptional modulator; TLE family member 5; amino-terminal enhancer of split; TLE5
Rabbit
Antigen affinity chromatography
RUO
166
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin)
4.235 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q08117
AES, TLE5
Recombinant Human Amino-terminal enhancer of split protein (1-264AA).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.