missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AES Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565563
This item is not returnable.
View return policy
Description
Immunogen sequence: MCHKNGFPQEG GITAAFLQKR KLRLSKNHRP ARAKVTEHVR GTRPGRATAG PAASTRAAGS LFFDRWGNRG PAGCRGSSHL PQQLKFTTSD Highest antigen sequence identity to the following orthologs - mouse 30%, rat 28%.
Adhesion to extracellular matrix regulates cell survival through both integrin engagement and appropriate cell spreading. Anoikis is the molecular mechanism of apop-tosis induced by integrin detachment. Amino-terminal enhancer of split (AES) is a member of the Groucho/ transducin-like enhancer of split (TLE) family of transcriptional regulators, a group of transcriptional co-repressors that play important roles in neurogenesis, segmentation, and sex determination. AES forms a complex with Bit1 (Bcl-2 inhibitor of transcription 1), a mitochondrial protein that is released into the cytoplasm upon onset of apoptosis. It has been suggested that this complex turns off a survival-promoting gene transcription program controlled by the TLE protein family. Interestingly, apoptosis of cells transfected with AES and Bit1 could be inhibited if the cells were allowed to attach to fibronectin through the alpha5beta1 integrin suggesting that the Bit1-AES pathway contributing to anoikis is regulated by integrins, and in particular, the alpha5beta1 integrin.
Specifications
| AES | |
| Polyclonal | |
| Unconjugated | |
| TLE5 | |
| Aes; AES-1; AES-2; AL024115; Amino enhancer of split; amino-terminal enhancer of split; Esp1; gp130-associated protein GAM; Grg; Grg5; Grg-5; Groucho-related protein 5; Protein ESP1; Protein GRG; related to Drosophila groucho; R-esp1; TLE family member 5; TLE family member 5, transcriptional modulator; TLE family member 5; amino-terminal enhancer of split; TLE5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 166 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q08117 | |
| AES, TLE5 | |
| Recombinant Human AES. Recombinant protein control fragment (Product #RP-106236). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction