Learn More
B Braun™ Aesculap™ Safety Scalpel
Description
- Safety locking mechanism for disposal
- Single-handed operation
- Ergonomic guide for precise incision
- Ruler in cm and inches
- Transparent cap for blade protection – blade and position is always visible
- Provides optimal sharpness
Specifications
Specifications
| Blade Material | Carbon Steel |
| Size | # 20 |
| Description | Surgeon Grade |
| Disposable | Single-use |
| Sterility | Sterile |
| Includes | Protective Cap |
| Handle Material | Plastic |
| Quantity | 10/Pk. |