Learn More
AF332 EP Color Scale
Used to assess product color by comparison with representative series of fixed color standards
Brand: Lovibond Tintometer 433320
Additional Details : Weight : 2.47000kg
Description
- Based on Lovibond Comparator System 2000, this kit is easy means of ordering complete range of equipment required for grading pharmaceutical solutions according to the EP Colour series
- Includes five colour-grading discs
- Each containing stable-coloured glass standards that match reference solutions specified in European Pharmacopoeia
- Sample (in 40mm glass cell) is viewed in Lovibond Comparator 2000+ with appropriate test disc until colour match is obtained
Specifications
Lovibond™ Comparator System 2000, five color-grading discs and two 40 mm cells | |
Comparator 2000+ |