missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF4 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28728G
This item is not returnable.
View return policy
Description
AF4 Polyclonal antibody specifically detects AF4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| AF4 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AF-4, AF4/FMR2 family member 1, AF4/FMR2 family, member 1, AF4myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 2, ALL1-fused gene from chromosome 4 protein, FEL, MLLT2MGC134969, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 2, PBM1, pre-B-cell monocytic leukemia partner 1, Protein AF-4, Protein FEL, Proto-oncogene AF4 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human proto-oncogene AF4 using the numbering given in entry NP_005926.1 (GeneID 4299). | |
| 0.1 mL | |
| Cancer | |
| 4299 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction