missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Afadin/AF-6 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-22996UV
This item is not returnable.
View return policy
Description
Afadin/AF-6 Polyclonal antibody specifically detects Afadin/AF-6 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Afadin/AF-6 | |
| Polyclonal | |
| Western Blot | |
| AF64, afadin, FLJ34371, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 4 | |
| The immunogen recognized by this antibody maps to a region between residue 1475 and 1525 of human AF6 (myeloid/lymphoid or mixed-lineage leukemia translocated to, 4 ) using the numbering given in entry BAA32484.1 (GeneID 4301). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4301 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction