missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Afadin/AF-6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 540.75
Specifications
| Antigen | Afadin/AF-6 |
|---|---|
| Concentration | 0.7mg/mL |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18443322
|
Novus Biologicals
NBP1-90219-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18253697
|
Novus Biologicals
NBP1-90219 |
0.1 mL |
€ 572.00 € 540.75 / 0.10mL Save € 31.25 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Afadin/AF-6 Polyclonal specifically detects Afadin/AF-6 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Afadin/AF-6 | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P55196 | |
| 4301 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ASHVFKFVDPSQDHALAKRSVDGGLMVKGPRHKPGIVQETTFDLGGDIHSGTALPTSKSTTRLDSDRVSSASSTAERGMVKPMIRVEQQPDYRRQESRTQD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.7mg/mL | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| AF64, afadin, FLJ34371, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 4 | |
| MLLT4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title