missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Afamin Polyclonal Antibody
GREENER_CHOICE

Product Code. 17149273
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17149273 100 μL 100µL
17178743 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17149273 Supplier Invitrogen™ Supplier No. PA5111355

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EMEKLVKDMV EYKDRCMADK TLPECSKLPN NVLQEKICAM EGLPQKHNFS HCCSKVDAQR RLCFFYNKKS DVGFLPPFPT LDPEEKCQAY ESNRESLLNH FLYEVARRNP FVFAPTLLTV AVHFEEVAKS CCEEQNKVNC LQTRAIPV Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 68%.

This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Afamin
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AFM
Gene Accession No. P43652
Gene Alias Afamin; AFM; ALB2; ALBA; Alf; alpha albumin; alpha-Alb; Alpha-albumin; MGC125338; MGC125339
Gene Symbols AFM
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Afamin. Recombinant protein control fragment (Product #RP-108395).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 173
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.