Learn More
Invitrogen™ Afamin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110823
Description
Immunogen sequence: EMEKLVKDMV EYKDRCMADK TLPECSKLPN NVLQEKICAM EGLPQKHNFS HCCSKVDAQR RLCFFYNKKS DVGFLPPFPT LDPEEKCQAY ESNRESLLNH FLYEVARRNP FVFAPTLLTV AVHFEEVAKS CCEEQNKVNC LQTRAIPV Highest antigen sequence indentity to the following orthologs: Rat - 68%, Mouse - 68%.
This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.
Specifications
| Afamin | |
| Polyclonal | |
| Unconjugated | |
| AFM | |
| Afamin; AFM; ALB2; ALBA; Alf; alpha albumin; alpha-Alb; Alpha-albumin; MGC125338; MGC125339 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 173 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P43652 | |
| AFM | |
| Recombinant Protein corresonding to Human Afamin. Recombinant protein control fragment (Product #RP-102380). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.