missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFAP Antibody (16F50), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22004AF647
This item is not returnable.
View return policy
Description
AFAP Monoclonal specifically detects AFAP in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
| AFAP | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Q12884 | |
| AFAP1 | |
| Human recombinant AFAP | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 16F50 | |
| Flow Cytometry, ELISA | |
| actin filament associated protein 1, FLJ56849 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 60312 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction