missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFAP Antibody (16F50), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22004MFV610
This item is not returnable.
View return policy
Description
AFAP Monoclonal antibody specifically detects AFAP in Human samples. It is validated for Flow Cytometry
Specifications
| AFAP | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 16F50 | |
| Flow Cytometry | |
| actin filament associated protein 1, FLJ56849 | |
| Human recombinant AFAP | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 60312 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction