missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFAP Antibody (OTI2G9), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72120APC
This item is not returnable.
View return policy
Description
AFAP Monoclonal antibody specifically detects AFAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| AFAP | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2G9 | |
| Western Blot, Immunohistochemistry | |
| actin filament associated protein 1, FLJ56849 | |
| Human recombinant protein fragment corresponding to amino acids 100-386 of human AFAP1 (NP_940997) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 60312 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction