missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFAP Antibody (OTI2G9), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72120C
This item is not returnable.
View return policy
Description
AFAP Monoclonal specifically detects AFAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| AFAP | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AFAP1 | |
| Recombinant protein fragment corresponding to amino acids 100-386 of human AFAP (NP_940997) produced in E.coli. | |
| 0.1 mL | |
| 60312.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2G9 | |
| Western Blot, Immunohistochemistry | |
| actin filament associated protein 1, FLJ56849 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction