missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFAP1 Monoclonal Antibody (OTI2A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810438S
This item is not returnable.
View return policy
Description
AFAP1 Monoclonal antibody specifically detects AFAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| AFAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N556 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 60312 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A3 | |
| Unconjugated | |
| Afap1 | |
| 110 kDa actin filament-associated protein; 2600003E23Rik; 9630044L16Rik; actin filament associated protein 1; actin filament-associated protein 1; actin filament-associated protein, 110 kDa; AFAP; Afap1; AFAP110; AFAP-110; AI848729; FLJ56849; im:7148276; Kiaa3018; mKIAA3018 | |
| Full length human recombinant protein of human AFAP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction