missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFAP1L2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92280-0.02ml
This item is not returnable.
View return policy
Description
AFAP1L2 Polyclonal antibody specifically detects AFAP1L2 in Human samples. It is validated for Western Blot
Specifications
| AFAP1L2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| actin filament associated protein 1-like 2, actin filament-associated protein 1-like 2, AFAP1-like protein 2, FLJ14564, KIAA1914Em:AC005383.4, XB130CTB-1144G6.4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human AFAP1L2 (NP_001001936.1). MERYKALEQLLTELDDFLKILDQENLSSTALVKKSCLAELLRLYTKSSSSDEEYIYMNKVTINKQQNAESQGKAPEEQGLLPNGEPSQHSSAPQKSLPDLPPPKMIPERKQLAIPKTESPEGYYEEAEPYDTSLNEDGEAVSSSYESYDEEDGSKGKSAPYQWPSPEAGIELMRDARICAFLWRKKWLGQWAKQLCVIKDNRLLCYKSSKDHSPQLDVNLLGSSVIHKEKQVRKKEHKLKITPMNADVIVLGLQSKDQAEQWLRVIQEVSGLPSEGASEGNQYTPDAQRFNCQKPDIAEK | |
| 0.02 mL | |
| Cancer, Signal Transduction | |
| 84632 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction