missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFAP1L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06316-50ul
This item is not returnable.
View return policy
Description
AFAP1L2 Polyclonal antibody specifically detects AFAP1L2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| AFAP1L2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5ul/mg of lysate | |
| actin filament associated protein 1-like 2, actin filament-associated protein 1-like 2, AFAP1-like protein 2, FLJ14564, KIAA1914Em:AC005383.4, XB130CTB-1144G6.4 | |
| Produced in rabbits immunized with E. coli-derived Human AFAP1L2 fragment. | |
| 50 μg | |
| Cancer, Signal Transduction | |
| 84632 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction