Learn More
AffiniPure Goat anti-Mouse, Biotin, Polyclonal, Proteintech
Goat Polyclonal Antibody
Brand: Proteintech SA00004-1-200UL
This item is not returnable.
View return policy
Description
AffiniPure Polyclonal antibody specifically detects AffiniPure in Human, Mouse samples. It is validated for ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| AffiniPure | |
| Polyclonal | |
| Biotin | |
| 200 μL | |
| Human, Mouse | |
| IgG |
| ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 0.2 mg/mL | |
| Goat | |
| Secondary | |
| Liquid |