Learn More
AffiniPure Goat anti-Mouse, HRP, Polyclonal, Proteintech
Goat Polyclonal Antibody
Brand: Proteintech SA00001-1-200UL
Additional Details : Weight : 0.00010kg
Description
AffiniPure Polyclonal antibody specifically detects AffiniPure in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot.
Specifications
AffiniPure | |
Polyclonal | |
HRP | |
200 μL | |
Human, Mouse, Rat | |
IgG |
ELISA, Western Blot | |
0.2 mg/mL | |
Goat | |
Secondary | |
Liquid |