missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFG3L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35543-20ul
This item is not returnable.
View return policy
Description
AFG3L2 Polyclonal antibody specifically detects AFG3L2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| AFG3L2 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA, Immunohistochemistry, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| AFG3 (ATPase family gene 3, yeast)-like 2, AFG3 ATPase family gene 3-like 2 (S. cerevisiae), AFG3 ATPase family gene 3-like 2 (yeast), AFG3-like protein 2, ATPase family gene 3, yeast, EC 3.4.24, EC 3.4.24.-, FLJ25993, Paraplegin-like protein, SCA28, spinocerebellar ataxia 28 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 538-797 of human AFG3L2 (NP_006787.2).,, Sequence:, DSINQKHFEQAIERVIGGLEKKTQVLQPEEKKTVAYHEAGHAVAGWYLEHADPLLKVSIIPRGKGLGYAQYLPKEQYLYTKEQLLDRMCMTLGGRVSEEIFFGRITTGAQDDLRKVTQSAYAQIVQFGMNEKVGQISFDLPRQGDMVLEKPYSEATARLIDDEVRILINDAYKRTVALLTEKKADVEKVALLLLEKEVLDKNDMVELLGPRPFAEKSTYEEFVEGTGSLDEDTSLPEGLKDWNKEREKEKEEPPGEKVAN | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10939 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction