missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aflatoxin Antibody (ATB), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-8086AF700
This item is not returnable.
View return policy
Description
Aflatoxin Monoclonal antibody specifically detects Aflatoxin in Nonspecies specific samples. It is validated for ELISA
Specifications
| Aflatoxin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| AFB1-AR1, Aflatoxin M1, AFT, Aldoketoreductase 7, FB1 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with aflatoxin. | |
| 0.2 mL | |
| Amino Acids Drugs and other small molecules | |
| Non-species specific | |
| Purified |
| ELISA | |
| ATB | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction