missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AFM Polyclonal Antibody
GREENER_CHOICE

Product Code. 17117303
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17117303 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17117303 Supplier Invitrogen™ Supplier No. PA5109367

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EDVSSNYDGC CEGDVVQCIR DTSKVMNHIC SKQDSISSKI KECCEKKIPE RGQCIINSNK DDRPKDLSLR EGKFTDSENV CQERDADPDT FFAKFTFEYS RRHPDLSIPE LLRIVQIYKD LLRNCCNTEN PPGCYRYAED KFNETTEKSL KMVQQECKHF QNLGKDGLKY HYLIRLTKIA PQLSTEELVS LGEKMVTAFT TCCTLSEEFA CVDNLADLVF GELCGVNENR TINPAVDHCC KTNFAFRRPC FESLKADKTY VPPPFSQDLF TFHADMCQSQ NEELQRKTDR FLVNLVKLKH ELTDEELQSL FTNFANVVDK CCKAESPEVC FNEESPKIGN.

This gene is a member of the albumin gene family, which is comprised of four genes that localize to chromosome 4 in a tandem arrangement. These four genes encode structurally-related serum transport proteins that are known to be evolutionarily related. The protein encoded by this gene is regulated developmentally, expressed in the liver and secreted into the bloodstream.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AFM
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AFM
Gene Accession No. P43652
Gene Alias Afamin; AFM; ALB2; ALBA; Alf; alpha albumin; alpha-Alb; Alpha-albumin; MGC125338; MGC125339
Gene Symbols AFM
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 260-599 of human AFM (NP_001124.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 173
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.