missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AFMID Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554901
This item is not returnable.
View return policy
Description
Immunogen sequence: VSGVGFPSKV PWKKMSAEEL ENQYCPSRWV VRLGAEEALR TYSQIGIEAT TRARATRKSL LHVPYGD Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 48%.
Catalyzes the hydrolysis of N-formyl-L-kynurenine to L-kynurenine, the second step in the kynurenine pathway of tryptophan degradation. Kynurenine may be further oxidized to nicotinic acid, NAD(H) and NADP(H). Required for elimination of toxic metabolites.
Specifications
| AFMID | |
| Polyclonal | |
| Unconjugated | |
| AFMID | |
| 9030621K19Rik; AFMID; Ammd; Arylformamidase; FKF; formylase; formylkynureninase; hypothetical protein LOC550372; im:7148587; Kf; KFA; KFase; kynurenine formamidase; N-formylkynurenine formamidase; probable arylformamidase; zgc:112472 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 125061 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q63HM1 | |
| AFMID | |
| Recombinant protein corresponding to Human AFMID. Recombinant protein control fragment (Product #RP-93517). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction