missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AFMID Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100379
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total AFMID.
Catalyzes the hydrolysis of N-formyl-L-kynurenine to L-kynurenine, the second step in the kynurenine pathway of tryptophan degradation. Kynurenine may be further oxidized to nicotinic acid, NAD(H) and NADP(H). Required for elimination of toxic metabolites.
Specifications
| AFMID | |
| Polyclonal | |
| Unconjugated | |
| AFMID | |
| 9030621K19Rik; AFMID; Ammd; Arylformamidase; FKF; formylase; formylkynureninase; hypothetical protein LOC550372; im:7148587; Kf; KFA; KFase; kynurenine formamidase; N-formylkynurenine formamidase; probable arylformamidase; zgc:112472 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 125061, 71562 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q63HM1, Q8K4H1 | |
| AFMID | |
| A synthesized peptide derived from human AFMID(Accession Q63HM1), corresponding to amino acid residues T142-L192. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction