missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFP Monoclonal Antibody (OTI4G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809082S
This item is not returnable.
View return policy
Description
AFP Monoclonal antibody specifically detects AFP in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| AFP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P02771 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 174 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunocytochemistry, Western Blot | |
| OTI4G9 | |
| Unconjugated | |
| AFP | |
| a fetoglobulin; a fetoprotein; Afp; AFPD; Alpha fetoglobulin; alpha fetoprotein; alpha-1-fetoprotein; Alpha-fetoglobulin; alpha-fetoprotein; alpha-fetoprotein precursor; alpha-foetoprotein; chloride intracellular channel 6; FETA; fetoglobulin; fetoprotein; HPAFP; a fetoglobulin; a fetoprotein | |
| Human recombinant protein fragment corresponding to amino acids 19-397 of human AFP produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction