missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AG-2/AGR2 Antibody (10E2), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27175AF647
This item is not returnable.
View return policy
Description
AG-2/AGR2 Monoclonal specifically detects AG-2/AGR2 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AG-2/AGR2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| O95994 | |
| AGR2 | |
| A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| 10551 | |
| Human, Mouse, Bovine, Canine, Equine, Other, Primate | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1000 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Primate/Monkey, Canine/Dog, Mouse, Bovine, Horse, Orangutan, Panda and Platypus. Immunogen sequence similarity with other species: Xenopus (82%), Chicken (83%). | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction