missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AG-2/AGR2 Antibody (10E2) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-27175CL1
This item is not returnable.
View return policy
Description
AG-2/AGR2 Monoclonal antibody specifically detects AG-2/AGR2 in Bovine,Canine,Equine,Human,Mouse,Primate samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| AG-2/AGR2 | |
| Monoclonal | |
| CoraFluor 1 | |
| AG-2, AG2member 17, anterior gradient 2 homolog (Xenopus laevis), anterior gradient homolog 2 (Xenopus laevis), GOB-4, hAG-2, HPC8, secreted cement gland homolog | |
| A portion of amino acids 50-100 of human AGR2 was used as the immunogen for the antibody. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| 10551 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 10E2 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Equine, Human, Mouse, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction