missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AG-3/AGR3 Antibody (AGR3.1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-45004AF488
This item is not returnable.
View return policy
Description
AG-3/AGR3 Monoclonal antibody specifically detects AG-3/AGR3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| AG-3/AGR3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| AG-3, anterior gradient homolog 3 (Xenopus laevis), BCMP11AG3, Breast cancer membrane protein 11, hAG-3member 18 | |
| Purified human AGR3 protein | |
| 0.1 mL | |
| Cancer | |
| 155465 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| AGR3.1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction