missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 50 μg/g in 75 cSt Hydrocarbon Oil, VHG™
Click to view available options
Quantity:
100 g
200 g
400 g
Unit Size:
100g
200g
400g
Description
Ag, Al, B, Ba, Bi, Ca, Cd, Cr, Cu, Fe, In, Li, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn @ 50 μg/g in 75 cSt Hydrocarbon Oil
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Copper/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-36-0, 7440-39-3, 7440-69-9, 7440-42-8, 7440-43-9 |
| Methods | ICP/XRF/RDE |
| Analyte or Component Names | Aluminium/Antimony/Barium/Bismuth/Boron/Cadmium/Calcium/Chromium/Copper/Indium/Iron/Lead/Lithium/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Hydrocarbon Oil |
| Quantity | 400 g |
| Concentration or Composition (by Analyte or Components) | Aluminium 50 μg/g/Antimony 50 μg/g/Barium 50 μg/g/Bismuth 50 μg/g/Boron 50 μg/g/Cadmium 50 μg/g/Calcium 50 μg/g/Chromium 50 μg/g/Copper 50 μg/g/Indium 50 μg/g/Iron 50 μg/g/Lead 50 μg/g/Lithium 50 μg/g/Magnesium 50 μg/g/Manganese 50 μg/g/Molybdenum 50 μg/g/Nickel 50 μg/g/Phosphorus 50 μg/g/Potassium 50 μg/g/Silicon 50 μg/g/Silver 50 μg/g/Sodium 50 μg/g/Tin 50 μg/g/Titanium 50 μg/g/Vanadium 50 μg/g/Zinc 50 μg/g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction