Learn More
AG14361, MedChemExpress
Product Code. 16481177
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16481177
Brand: Medchem Express HY12032
MedChemExpress AG14361 is a potent PARP-1 inhibitor, with a Ki of IC50s are 29 nM and 14 nM, respectively.
AG14361 is a potent PARP-1 inhibitor, with a Ki of IC50s are 29 nM and 14 nM, respectively.
Specifications
Chemical Name or Material AG14361
CAS 328543-09-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H20N4O
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (156.06 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1NCCN2C3=C1C=CC=C3N=C2C4=CC=C(C=C4)CN(C)C
Molecular Weight (g/mol) 320.39
Formula Weight 320.39
Percent Purity 99.05%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only