Learn More
Invitrogen™ AGAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550542
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Centaurin gamma 2 protein.
GTPase-activating protein for ARF1 and, to a lesser extent, ARF5. Directly and specifically regulates the adapter protein 3 (AP-3)-dependent trafficking of proteins in the endosomal-lysosomal system.
Specifications
| AGAP1 | |
| Polyclonal | |
| Unconjugated | |
| AGAP1 | |
| AGAP1; AGAP-1; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 1; ArfGAP with GTPase domain, ankyrin repeat and PH domain 1; arf-GAP with GTPase, ANK repeat and PH domain-containing protein 1; centaurin, gamma 2; centaurin-gamma-2; CENTG2; cnt-g2; GGAP1; GTP-binding and GTPase-activating protein 1; Kiaa1099; mKIAA1099; R75364 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116987, 347722 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8BXK8, Q9UPQ3 | |
| AGAP1 | |
| Synthetic peptide corresponding to a region derived from internal residues of human ArfGAP with GTPase domain, ankyrin repeat and PH domain 1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |