Learn More
Melford Agar Bacteriological Grade
Product Code. 30017964
Click to view available options
:
100 g
Product Code. 30017964
Brand: Melford A20030100.0
To Prepare: Mix 15 grams per liter in Molecular Biology grade water.
To Prepare: Mix 15 grams per liter in Molecular Biology grade water.
Specifications
Quantity 100 g