Learn More
Melford Agar Bacteriological Grade
Click to view available options
:
100 g
Description
To Prepare: Mix 15 grams per liter in Molecular Biology grade water.
To Prepare: Mix 15 grams per liter in Molecular Biology grade water.
Specifications
Specifications
| Quantity | 100 g |