Learn More
RETSCH Agate Grinding Jars
Shop All Retsch Products
Click to view available options
For Use With (Equipment):
For MM 200 Mixer Mill
For MM 301 Mixer mill
Capacity (Metric):
5 mL
10 mL
Unit Size:
Each
Description
- Screw-top grinding jars
- Available in a variety of sizes
- For use with Retsch mills
Specifications
Specifications
| Product Type | Grinding Jar |
| Capacity (Metric) | 5 mL |
| For Use With (Equipment) | For MM 301 Mixer mill |
| Material | Agate |