Learn More
RETSCH Agate Hand Mortar
Brand:  RETSCH 20.262.0001
Additional Details : Weight : 0.07000kg
Product Code. 10044831
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
8 mL
40mm
Agate
Hand Mortar
30mm
Hand Mortar
Suitable for grinding, mixing and triturating medium-hard to hard substances