Learn More
Beckman Coulter™ Agencourt AMPure XP
Click to view available options
Volume:
5mL
60mL
450mL
Unit Size:
450mL
Each
Description
- SPRI paramagnetic bead-based chemistry
- Removes contaminants (dNTPs, salts, primers, primer dimers)
- High recovery of amplicons (>100 bp) ensures that critical data is not lost
- Trusted and recommended reagent for a variety of NGS workflows
- Compatible with manual and automated processing
- Predictable and consistent size selection
- Manual cleanups using AMPure XP require less than 30 minutes
- AMPure XP is automated on all Biomek platforms for higher throughput
Specifications
Specifications
| Composition | AMPure XP, Speed Beads (Mag Carboxy) |
| Content And Storage | 2-8°C |
| Form | Liquid |
| For Use With (Equipment) | Automated 96- or 384-well formats, e.g. Biomek platforms |
| pH | 8.0 to 8.4 |
| For Use With (Application) | Standard PCR, Real Time PCR (qPCR), Next-Generation Sequencing (NGS) |
| Sample Type | DNA |
| Volume | 450mL |