Learn More
AGER Antibody (697023) [Janelia Fluor™ 646], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB11795J
Product Code. 18538246
  • € 578.00 / 0.10mL
Description
AGER Monoclonal antibody specifically detects AGER in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AGER
Monoclonal
Janelia Fluor 646
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Western Blot, Flow Cytometry, CyTOF
697023
Western Blot, Flow Cytometry, CyTOF-ready
advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products
Mouse myeloma cell line NS0-derived recombinant mouse RAGE, Gly23-Leu342, Accession # NP_031451
0.1 mL
Cancer
177
Store at 4°C in the dark.