missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGER Antibody (CL10587), Novus Biologicals™
Mouse Monoclonal Antibody
€ 307.00 - € 538.00
Specifications
| Antigen | AGER |
|---|---|
| Clone | CL10587 |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649248
|
Novus Biologicals
NBP3-07982-25ul |
25 μg |
€ 307.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18606178
|
Novus Biologicals
NBP3-07982-100ul |
100 μg |
€ 538.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGER Monoclonal antibody specifically detects AGER in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| AGER | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: VSEGSYPAGTLSWHLDGK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL10587 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS, pH 7.2, containing 40% glycerol | |
| 177 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title