missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG F(ab')2 Secondary Antibody, DyLight 650, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-61921C
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Human IgG F(ab')2 fragments | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| DyLight 650 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction