missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aggrecan Neoepitope Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74350PCP
This item is not returnable.
View return policy
Description
Aggrecan Neoepitope Polyclonal antibody specifically detects Aggrecan Neoepitope in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Flow Cytometry
Specifications
| Aggrecan Neoepitope | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Canine | |
| Purified |
| Flow Cytometry | |
| PerCP | |
| AGC1SEDK, aggrecan, aggrecan core protein, Cartilage-specific proteoglycan core protein, Chondroitin sulfate proteoglycan 1, Chondroitin sulfate proteoglycan core protein 1, CSPCP, CSPG1aggrecan 1, CSPGCP, large aggregating proteoglycan, MSK16AGCAN | |
| A synthetic peptide corresponding to a region of human Aggrecan (within residues 350-400). [UniProt# P16112] | |
| 0.1 mL | |
| Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 176 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction