Learn More
Brady™ Aggressive Adhesive, Multipurpose Clear Polyester Labels
Label circuit boards, panels and equipment with the B-432 clear polyester label material, allowing the applied surface colors to transmit around the printed label text. Print almost anywhere with a Brady™ handheld label maker.
€ 62.08 - € 64.70
Specifications
| Certifications/Compliance | UL Recognised;CSA Accepted Compliant with Directive 2011/65/EU and 2015/863 |
|---|---|
| Adhesive Type | Permanent Acrylic |
| Color | Transparent, Clear |
| Finish | Gloss |
| Format | Die-cut |
Description
- Ideal for electronic PCB and component identification, barcode labels, rating plates and solar panel identification
- Made from tough clear polyester that's chemical, oil, water and extreme temperature resistant
- Features a permanent acrylic adhesive that's ideal for smooth and rough surfaces alike
Specifications
| UL Recognised;CSA Accepted Compliant with Directive 2011/65/EU and 2015/863 | |
| Transparent, Clear | |
| Die-cut | |
| Non-sterile | |
| BMP51;BMP53;M511;M510 | |
| -40°C to 100°C | |
| 0.102 mm |
| Permanent Acrylic | |
| Gloss | |
| Polyester | |
| Asset tracking;Barcode labelling;Component and equipment labelling;Data and telecommunications labelling;Electrical labelling;General identification;Inventory and inspection labelling;Warehouse marking | |
| -40°F to 212°F | |
| 0.004 in | |
| 1 in |