Learn More
STEMCELL Technologies AggreWell™800 24-well Plate, 1 pack
Product Code. 15820967
Encompass_Preferred
Click to view available options
Quantity:
1 Plate
Unit Size:
Each
Product Code. 15820967
Brand: STEMCELL Technologies 34811
AggreWell™800 (1 x 24-well plate)
  • AggreWell™ plates bring an easy, standardized approach to the generation of cell aggregates, including embryoid bodies (EBs) and spheroids
  • EBs and spheroids generated using AggreWell™ plates are consistent in size and shape, and are uniform within and between experiments
  • New and improved second-generation AggreWell™ plates are compatible with a variety of cell types, including ES and iPS cells, cancer cells and more
  • Enhanced optical characteristics provide crystal clear imaging
  • Note: AggreWell™ Rinsing Solution is required for optimal EB and spheroid formation
Specifications
For Use With (Application) Differentiation; Spheroid Culture; Toxicity Assay
Quantity 1 Plate